General purpose gloves, Showa 341
Supplier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
112-6003EA 80.2 GBP
112-6003 112-6004 112-6005 112-6006
General purpose gloves, Showa 341
Gloves
These nylon/polyester knitted gloves are hard-wearing and highly tactile, even in wet environments.
  • Natural rubber coating on the palm is water-resistant
  • Rough surface grip enables optimum dexterity when working with small components or heavy components
  • Elasticated wrist cuff prevents dirt and debris from entering the glove
  • Seamless knit offers ultimate comfort during long periods of wear
Typical applications include: Decoration, drainage, piping, flashings and seals, guttering and drain pipe, electrical wiring, insulation, roofing application membranes.
Certifications: EN 388:2016 (2121X); Cat. II
Packaging: 10 pairs per polybag.