Imprimer
Gas-safety adapter 360 for Flame 100
Fournisseur: WLD-Tec
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WART2.000.701EA 154 EUR
WART2.000.701
Gas-safety adapter 360 for Flame 100
Appareils chauffants Brûleurs à gaz
Gas-safety adapter 360 for Flame 100.
  • With integrated pressure regulator
  • Continuous cartridge operation 65 minutes
Ideal for cartridges CV 360, 52 g from Camping gas.