Imprimer
Signs, COVID-19 General safety information, PP, Brady®
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BRDY306944EA 25 EUR
BRDY306944
Signs, COVID-19 General safety information, PP, Brady®
Signs are made of surface printed rigid polypropylene material with a slightly textured surface. They are overlaminated with a gloss clear polyester.
  • Ideal for indoor and outdoor applications
  • Well suited for harsh environments
  • Good solvent resistance
  • Rectangular shape
  • Colours: Black and white on red, green, blue, yellow and white
Thickness: 1,501 mm.