Imprimer
Industrial gloves, AlphaTec® 87-063
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AlphaTec®
ANSE87-063/7.5EA 58.8 EUR
ANSE87-063/7.5 ANSE87-063/8.5 ANSE87-063/9.5 ANSE87-063/10.5
Industrial gloves, AlphaTec® 87-063
Gants
Flocked natural rubber latex gloves with mechanical and chemical resistance.
  • 100% cotton flock lining for better comfort
  • High resistance to water based chemicals
  • Raised pattern on palms and fingers for excellent dry and wet grip
  • Longer cuff for extra protection
  • Beaded cuff provides tear resistance and allows easy donning
These flexible and comfortable gloves offer good abrasion resistance.
Recommended for: Chemical handling, food processing applications and janitorial use.
Certifications: EN ISO 374-1:2016 (AKLNPT), EN ISO 374-5 VIRUS; EN 388:2016 (3110A); Cat. III; AQL 0,65
Food contact approved
Cdt: 12 pairs per bag.