Imprimer
Industrial label printers with Brady workstation LAB suite, BradyPrinter i5100
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BRDY306872EA 1456 EUR
BRDY306872 BRDY306876 BRDY306874 BRDY306878
Industrial label printers with Brady workstation LAB suite, BradyPrinter i5100
Imprimantes Label Printers
The BradyPrinter i5100 Industrial Label Printer offers a perfect mix of performance and smarts.
  • 300 or 600 dpi
  • Mono colour
  • Applications include asset identification, circuit board and component identification, and general and industrial identification
This benchtop printer can print on 70 label materials, more than 1200 stock label parts and tailored custom label materials and parts. Applications include a wide range of specialised and commonly used product labels, electronics and PCB labels, logistics and asset tracking labels, cable & component identification labels and laboratory sample labels.
Materials supported include Direct Thermal Paper; Freezerbondz™ Polyester; Heat-shrink Polyolefin Sleeves; High Temperature Heat-shrink Polyvinylidene Fluoride Sleeves; Metallic, Non-metal Polyester; Metallised Polyester; Nylon Cloth; Paper; Polyester; Polyethylene Foam Laminate Polyester; Polyethylene Tag; Polyimide; Polymer; Polypropylene; Polypropylene Tag; Polyvinylfluoride; Removable Polyester; Repositionable Paper; Self-laminating Polyester; Self-laminating Vinyl; StainerBondz™ Polyester; Static Dissipative Polyester; Tamper-evident Metallised Polyester; Tamper-evident Polyester; Tamper-resistant Metallised Vinyl; Vinyl Cloth; Water-indicating Polyester.
Barcode types include Code 39; Code 93; Code 39 Full ASCII; code 128 A; code 128 B; code 128 C; EAN 8; EAN13; EAN/UCC 128/GS1-128; EAN/UPC; FIM; HIBC; Interleaved 2/5; ident and lead code of Deutsche Post AG; Codabar; JAN8; JAN 13; MSI; Plessey; Postnet; RSS14; UPC A; UPC E; UPC E0; Aztec; Codablock F; DataMatrix; PDF417; Micro PDF417; UPS MaxiCode; QR code; RSS 14 truncated, limited stacked and stacked omnidirectional; EAN/GS1 DataMatrix; GS1 DataBar.
Languages: Arabic (Egypt); Bulgarian; Chinese (Traditional and Simplified); Croatian; Czech; Danish; Dutch; English; Finnish; French; German; Greek; Hungarian; Italian; Lithuanian; Macedonian; Norwegian (Bokmål); Persian (Farsi); Polish; Portuguese; Romanian; Russian; Serbian; Slovenian; Spanish; Swedish; Turkish; Thai; Hebrew.
Informations sur les accessoires: Available hardware options include Ribbon Take up Spindle; Label Roll Holder; and Platen Rollers; contact your Avantor representative for more information.
Informations de livraison: This i5100 printer comes in a cardboard box. It ships with a power cord, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English and a printed user manual in English, an RFID enabled roll holder, a ribbon take up spindle and a Brady Workstation Laboratory Identification Suite.