Imprimer
ICP-OES Wavelength calibration solution
Fournisseur: Honeywell Research Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HONCE2009-100MLEA 644 EUR
HONCE2009-100ML
ICP-OES Wavelength calibration solution
Certified Reference Material (CRM).
50 mg/L K, 5 mg/L each of Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn in HNO₃ 5%