Imprimer
Face shields, disposable, GUARDALL SHIELD™
Fournisseur: Kimberly-Clark
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GUARDALL SHIELD™
KIMB41205EA 302 EUR
KIMB41205 KIMB41204
Face shields, disposable, GUARDALL SHIELD™
Écrans de protection
The full-length face shield is designed for procedures and environments where there’s a risk of exposure to bodily fluids and / or blood through splashing.
  • Equipped with a foam band and headband for comfort and easy donning
  • Can be worn with our without glasses or goggles
  • For use in humid environments or places with temperature variations (as well as anywhere you need this protection)
  • Clear visor for maximum visibility
It’s roomy enough to wear over glasses or goggles and is resistant to fogging. The clear visor allows maximum visibility for all lab tasks.