Peroxidase (Myeloperoxidase) Leukocyte Kit DAB substrate kit, Sigma-Aldrich®
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
M-Clarity™ quality level = MQ500
Used to demonstrate polymorphonuclear cells in blood or bone marrow films. Each kit has sufficient reagents for 10 uses.
- Suitable for haematology
- Suitable for histology
- Sufficient for 10-120 manual assays
Diaminobenzidine, as a benzidine substitute for myeloperoxidase, has less tinctorial power than benzidine. This made DAB much less attractive to histochemists. Hanker and associates improved DAB methodology. According to their modification, the brown reaction product is first intensified with copper salts and followed by application of Gill′s modified Papanicolaou stain, resulting in intense grey-black staining at sites of neutrophil and monocyte peroxidase.
Staining system for polymorphonucleocytes in blood or bone marrow films. The staining characteristics of polymorphonucleocytes are used to distinguish acute myelocytic leukemia from other types of leukemia.
Kit component only
Diaminobenzidine 10 vial(s)
Copper nitrate 2 vial(s)
Gill Modified EA solution 100 mL
Glutaraldehyde solution 2 x 75 mL
Hematoxylin Solution, Gill No. 3 100 mL
Scott's Tap Water Substitute Concentrate 100 mL
TRIZMAL™ Buffer Concentrate 50 mL
Température de stockage:
Réfrigérateur |
Numéro MDL:
MFCD00131641 UN: 3316 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...