Imprimer
Phloxine B, Sigma-Aldrich®
Marques : SIGMA-ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): CI 45410
P4030-25GEA 329 EUR
P4030-25G
Phloxine B, Sigma-Aldrich®
Phloxin B
Phloxine B has been used as a component of yeast extract peptone dextrose (YEPD) to grow Schizosaccharomyces pombe.