Erbium(III) oxalate hydrate 99,9%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
APOSIN1723-25GEA
219
GBP
APOSIN1723-25G
Erbium(III) oxalate hydrate 99,9%
Erbium(III) oxalate hydrate
Formula:
(C₂O₄)₃·Er₂·H₂O MW: 616.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150141 CAS Number: 58176-72-0 |