1-Benzoylpiperazine
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Phenyl(piperazin-1-yl)methanone
APOSOR0862-1GEA
14.7
GBP
APOSOR0862-1G
1-Benzoylpiperazine
1-Benzoylpiperazine
Formula:
C₁₁H₁₄N₂O MW: 190.24 g/mol Boiling Pt: 145 °C Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00810192 CAS Number: 13754-38-6 |