4-Butyl-4'-cyanobiphenyl 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4'-Butyl-[1,1'-biphenyl]-4-carbonitrile
APOSOR13992-25GEA
168
GBP
APOSOR13992-25G
4-Butyl-4'-cyanobiphenyl 98%
4-Butyl-4'-cyanobiphenyl
Formula:
C₁₇H₁₇N MW: 235.33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00506009 CAS Number: 52709-83-8 |