Tetrachloropicolinic acid 96%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3,4,5,6-Tetrachloropyridine-2-carboxylic acid
APOSOR2044-500GEA
0
GBP
APOSOR2044-500G
APOSOR2044-5G
APOSOR2044-25G
Tetrachloropicolinic acid 96%
Tetrachloropicolinic acid
Formula:
C₆HCl₄NO₂ MW: 260.89 g/mol Melting Pt: 171…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185833 CAS Number: 10469-09-7 EINECS: 233-956-4 |