2-Amino-N-sec-butylbenzamide
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
APOSOR21223-5GEA
0
GBP
APOSOR21223-5G
APOSOR21223-25G
2-Amino-N-sec-butylbenzamide
2-Amino-N-sec-butylbenzamide
Formula:
C₁₁H₁₆N₂O MW: 192.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00116303 CAS Number: 30391-87-8 |