2-Nitro-4-thiophenecarboxylic acid
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
5-Nitrothiophene-3-carboxylic acid
APOSOR24046-10GEA
0
GBP
APOSOR24046-10G
APOSOR24046-1G
APOSOR24046-5G
2-Nitro-4-thiophenecarboxylic acid
2-Nitro-4-thiophenecarboxylic acid
Formula:
C₅H₃NO₄S MW: 173.15 g/mol Melting Pt: 145…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00052582 CAS Number: 40357-96-8 |