Methyl-3-cyano-4-methoxybenzoate
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Methyl 3-cyano-4-methoxybenzoate
APOSOR30137-5GEA
350
GBP
APOSOR30137-5G
APOSOR30137-1G
Methyl-3-cyano-4-methoxybenzoate
Methyl 3-cyano-4-methoxybenzoate
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol Melting Pt: 124…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052930 CAS Number: 25978-74-9 UN: 3439 ADR: 6.1,III |