3-(tert-Butylamino)propionitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(tert-Butylamino)propanenitrile

APOSOR322043-5GEA 0 GBP
APOSOR322043-5G
3-(tert-Butylamino)propionitrile
3-(tert-Butylamino)propionitrile
Formula: C₇H₁₄N₂
MW: 126.1995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045637
CAS Number: 21539-53-7

Order Now

Learn more