Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
, Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate
APOSOR322284-5GEA
0
GBP
APOSOR322284-5G
APOSOR322284-1G
Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate
Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate
Formula:
C₇H₁₁N₃O₂ MW: 169.18 g/mol Melting Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD01210846 CAS Number: 23286-70-6 |