2-Methoxybiphenyl 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

APOSOR937140-25GEA 97.4 GBP
APOSOR937140-25G
2-Methoxybiphenyl 95%
2-Methoxybiphenyl
Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008367
CAS Number: 86-26-0

Order Now

Learn more