Imprimer
ViraPack transfection kit
Fournisseur: Agilent Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
STRA200488EA 0 EUR
STRA200488
ViraPack transfection kit
Réactifs de transfection
The ViraPack transfection kit is based on a modified CaPO₄ method of DNA transfection to introduce viral plasmids into HEK293 packaging cell lines with high efficiency.
  • Unprecedented efficiency with retroviral, adenoviral, and AAV systems
  • Outperforms liposome-mediated transfection reagents in HEK293 cells
Vector details: Transfection kit designed specifically for HEK293 cells used in packaging virus for viral gene delivery systems.