Lambda genomic vector kits
Supplier: Agilent Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AGLS247713EA 1597 GBP
AGLS247713 AGLS248712 STRA248211
Lambda genomic vector kits
Vectors, Plasmids and Libraries
Lambda DASH II and Lambda FIX II vectors offer traditional genomic cloning of 9 to 23 kB fragments.
These vectors are used for genomic library construction contain T3 and T7 RNA promoters for rapid gene walking and high-resolution gene mapping.
Caution: For research use only. Not for use in diagnostic procedures.