Imprimer
pBC phagemid vectors
Fournisseur: Agilent Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AGLS212218EA 580 EUR
AGLS212218 AGLS212217 AGLS212216 AGLS212215
pBC phagemid vectors
Vecteurs, plasmides et librairies
The pBC vectors were derived from the pBluescript II phagemid. The ampicillin-resistance gene has been replaced with the chloramphenicol resistance gene.
  • Convenient selection when subcloning from ampicillin-resistant cloning vectors
  • Large and versatile polylinker in two orientations for cloning flexibility
  • Available with f1 origin in either (+) or (–) orientation
pBC phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
Attention: For research use only. Not for use in diagnostic procedures.