Imprimer
MBS Mammalian Transfection kit
Fournisseur: Agilent Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AGLS200388EA 667 EUR
AGLS200388
MBS Mammalian Transfection kit
The MBS mammalian transfection kit can increase transfection efficiencies up to 100-fold.
  • Additionally, cells can be assayed 12 to 24 hours post-transfection instead of the usual 48 to 72 hours.
Cells are incubated with CaPO₄/DNA coprecipitate for only three hours, versus 6 to 12 hours with traditional CaPO₄ methods.
For highest viral titer use this kit to transfect HEK293 cells. Solution 1: 2,5 M CaCl₂, Solution 2: 2x BBS (pH 6,95), Solution 3: Modified bovine serum (MBS), Control Plasmid: pCMV, Beta-gal.
Attention: For research use only. Not for use in diagnostic procedures.