Imprimer
Votre recherche pour: Pièges à vide
18  résultat(s)
SearchResultCount:"18"
Sort Results
Vue liste Nouvelle Vue
Evaluer les résultats de votre recherche
Référence Produit: (7520-94)
Fournisseur: Avantor
Description: The SPE-12G PTFE column processors are specially designed PTFE vacuum chambers that allow 12 samples to be run simultaneously. This offers the flexibi...
UOM: 1 * 1 Pce
Fournisseur: Avantor
Description: Ces systèmes de traitement des colonnes en verre sont des chambres de vide en verre à la conception spéciale permettant de traiter de multiples échant...
Fournisseur: KIMA SAS DI CHIARIN RENZO & C
Description: ESA 1 single use holder, PP
Référence Produit: (BOHLB317-60)
Fournisseur: Bohlender
Description: PTFE head with two threaded necks GL14 for connection of hard-walled tubing by means of separately available laboratory screw joints. Connecting nut ...
UOM: 1 * 1 Pce
Référence Produit: (KGWA17410-NW16)
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
UOM: 1 * 1 Pce
Fournisseur: Lenz Laborglas GmbH & CO.KG
Description: For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the ...
promo
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glas...
Référence Produit: (KGWA1707)
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents and these cold traps can be used for m...
UOM: 1 * 1 Pce
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: These cold traps (cold finger condensers) are constructed in the same way as standard cold traps.
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: Type S cold trap has a conical ground joint NS 45/40 as connection part. The condensation area can be separated from the upper section of cold trap. T...
Référence Produit: (KGWA1702)
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: This cold trap is a complete version in which the dewar flask is bonded to the cold trap, because of its design, this version has got two free walls f...
UOM: 1 * 1 Pce
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents.
Référence Produit: (KGWA17380-NW16)
Fournisseur: KGW (KARLSRUHER GLASTECHN.)
Description: Simple cold traps, which are also known as cold finger condensers, are used in conjunction with vacuum pumps and the condensation Produced from humidi...
UOM: 1 * 1 Pce
Fournisseur: PerkinElmer
Description: Thermal desorber trap, TurboMatrix, packed with tenax
Référence Produit: (ALCA066889)
Fournisseur: ALCATEL
Description: Liquid nitrogen trap condenses all gases at the pump inlet whose critical condensation temperature is above –196 °C. It protects the pump against cond...
UOM: 1 * 1 Pce
Fournisseur: EDWARDS HIGH VACUUM (METTLER)
Description: Inlet Desiccant Trap IDT20K: Use when pumping limited quantities of water vapour at high pumpings speeds to a low vapour pressure.
Prix sur demande
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Ces articles ne peuvent pas être ajoutés à votre panier en raison des exigences réglementaires. Veuillez envoyer un e-mail à webshop.fr@vwr.com
Veuillez noter qu'une autorisation peut être nécessaire pour commander ce produit. Un représentant de VWR vous contactera si votre commande le nécessite.
Ce produit a été bloqué par votre entreprise. Veuillez s'il vous plait contacter votre service achats pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Ce produit n'est plus disponible. Des alternatives peuvent être disponibles en faisant une recherche avec la référence VWR ci-dessus. Si vous avez besoin d'aide, s'il vous plaît appelez le Service Clients au 0825 02 30 30 (0,18 € TTC/mn).
1 - 16 of 18
no targeter for Bottom