Imprimer
Votre recherche pour: Essais protéiniques
81  résultat(s)
SearchResultCount:"81"
Sort Results
Vue liste Nouvelle Vue
Evaluer les résultats de votre recherche
Fournisseur: SIGMA-ALDRICH MICROSCOPY
Description: Bicinchoninic acid is used for the determination of protein concentration. It is responsible for the making of intense purple complex with cuprous ion...
Référence Produit: (1B1301-KIT)
Fournisseur: VWR Chemicals
Description: The Protein Precipitation Kit utilises a simple trichloroacetic acid (TCA) based method to precipitate proteins in complex mixtures. Only protein in t...
UOM: 1 * 1 KIT
Référence Produit: (M172-1L)
Fournisseur: VWR Chemicals
Description: Le dosage de protéines Bradford est le dosage colorimétrique privilégié pour quantifier la concentration totale en protéines. Basée sur la formation c...
UOM: 1 * 1 L
Fournisseur: Rockland Immunochemicals
Description: Conjugate stabilisers are used with horseradish peroxidase (HRP) and calf intestinal alkaline phosphatase conjugated primary or secondary antibodies o...
Fournisseur: Enzo Life Sciences
Description: The EFLUXX-ID® Multidrug Resistance Assay Kit allows for functional detection of all three clinically relevant ABC transporter proteins MDR1 (p-glycop...
Fournisseur: G-Biosciences
Description: Protein assay, Neutralizer™
Fournisseur: Enzo Life Sciences
Description: PROTEOSTAT® Protein aggregation assay provides a simple, homogenous assay format for monitoring peptide and protein aggregation in solution. This is u...
Fournisseur: Biotium
Description: Kit for fluorescence-based measurement of protein unfolding or thermal stability in thermal shift assay, also called Protein Thermal Shift™, different...
Fournisseur: G-Biosciences
Description: These CL (Compatible Lowry) Protein Assay is based on the widely cited protein assay by Lowry et. al. (1951). The CL Protein Assay improves upon the t...
Fournisseur: G-Biosciences
Description: Bicinchoninic Acid (BCA) Protein Assay, 1000 assays or 5000 microassays (with BSA protein standard)
Référence Produit: (CAYM700510-96)
Fournisseur: Cayman Chemical
Description: A fluorescence-based method for detecting L-lactate in biological samples such as serum, plasma, blood, urine, and saliva.
UOM: 1 * 1 Pce
Fournisseur: G-Biosciences
Description: The Micro Bicinchoninic Acid Protein Assay is a highly sensitive colorimetric assay that is compatible with detergent solubilised protein solutions. T...
Fournisseur: G-Biosciences
Description: CB-X™ Protein Assay with albumin standard, 500 assays
Fournisseur: Enzo Life Sciences
Description: This kit has been designed for monitoring protein stability under systematic thermal stress conditions.
Référence Produit: (786-005)
Fournisseur: G-Biosciences
Description: A highly sensitive colorimetric protein assay that overcomes interference of common laboratory agents present in protein solutions and shows minimal p...
UOM: 1 * 1 KIT
Fournisseur: G-Biosciences
Description: Bicinchoninic Acid (BCA) Reducing Agent Compatible Protein Assay (with non-animal protein standard)
Prix sur demande
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Ces articles ne peuvent pas être ajoutés à votre panier en raison des exigences réglementaires. Veuillez envoyer un e-mail à webshop.fr@vwr.com
Veuillez noter qu'une autorisation peut être nécessaire pour commander ce produit. Un représentant de VWR vous contactera si votre commande le nécessite.
Ce produit a été bloqué par votre entreprise. Veuillez s'il vous plait contacter votre service achats pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Ce produit n'est plus disponible. Des alternatives peuvent être disponibles en faisant une recherche avec la référence VWR ci-dessus. Si vous avez besoin d'aide, s'il vous plaît appelez le Service Clients au 0825 02 30 30 (0,18 € TTC/mn).
1 - 16 of 81
no targeter for Bottom