Flexacam C3, a 12 MP microscope camera for simplicity, colour accuracy, and real-time collaboration.
- Camera simplicity without a PC
- Flexibly enhance documentation with intuitive measurement, annotation, overlay, and network tools
- View images directly on a monitor and re-cord videos in full HD with up to 30 frames per second
- Save images to a local network or send as email attachment for easy sharing and storage
- Powerful colour accuracy
- Precise colour reproduction is provided by the sensor‘s dynamic range and signal processing unit
- Fine details of the sample are revealed thanks to the 12 MP CMOS sensor
- Fast screening of sample with bright and dark areas due to the fast auto exposure
- Collaboration in real time
- Clear and smooth visualisation of sample movements on the screen thanks to the high frame rate of up to 60 fps and high signal transfer speed from sensor to monitor
- Viewing of live and captured images directly on a monitor simultaneously by multiple users when used in stand-alone or workstation mode
Save time when capturing, documenting, and sharing images with the Flexacam C3 microscope camera. It transforms your microscope into a stand-alone digital imaging station without the need for a PC. Make reliable and accurate decisions for your application tasks with powerful colour accuracy. True-to-life colours and fine details are a given thanks to the 12 MP CMOS sensor, large dynamic range, and fast signal processing.
Collaborate in real time and review images efficiently as a team by taking advantage of the fast display of live images.