Imprimer
Industrial safety helmet, EVOLite®
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
EVOLite®
JSPAAJB170-000-100EA 43.7 EUR
JSPAAJB170-000-100 JSPAAJB170-000-200 JSPAAJB170-000-500
Industrial safety helmet, EVOLite®
Casques Casques de sécurité
EVOLite® is one of the lightest safety helmets for sale in the UK, conforming to EN 397.
  • Lightweight industrial safety helmet
  • Tough ABS shell
  • Wheel ratchet adjustment
  • Chamlon sweatband
The EVOLite® Industrial Safety Helmet is lightweight, extremely comfortable, and fits the widest range of head shapes and sizes. The harness and shell are truly integrated as one to give the best performance by keeping it firmly rooted on the head, even when fully inverted. Gives added security and protection when working at height, in high winds, or when there is a high degree of activity, such as during motorway maintenance.
Weighing less than 300 g (dependent on model), the EVOLite® is the lightest safety helmet for sale in the UK. A 6-point terylene cradle harness system offers unrivalled comfort without compromising performance. Egyptian cotton core with porous PU coating for maximum sweat absorption. pH neutral, dermatologically tested. 3-D precision fitting uses the unique 1-2-3 point harness depth settings.
Certifications: EN 397.