Imprimer
Flow Control Spring Tubing Clamps, Acetal
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX06834-10EA 47.4 EUR
MFLX06834-10 MFLX06834-20
Flow Control Spring Tubing Clamps, Acetal
Pinces Clips pour tubes
An ideal option to secure tubing.
  • Constructed of durable acetal copolymer and stainless steel
Acetal copolymer and stainless steel clamps provide on/off flow control for flexible tubing. Chemically resistant. Bel-Art. 12 bg/cs.