Imprimer
Masterflex® Modular Touch-Screen Dispensing Pump, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Masterflex®
MFLB07582-20EA 4179.06 EUR
MFLA07582-20 MFLX07582-20-IL MFLX07582-10-UK MFLX07582-20-CH MFLX07582-20-IN MFLB07582-20 MFLX07582-00-USS MFLX07582-00-DK MFLX07582-20-AU MFLX07582-10-DK MFLX07582-00-UK MFLX07582-20-IT MFLX07582-20-EU MFLX07582-00 MFLX07582-10-AU MFLX07582-00-IT MFLX07582-00-IN MFLX07582-20-DK MFLX07582-00-CH MFLX07582-00-IL MFLX07582-20-USS MFLX07582-10-EU MFLX07582-10-IN MFLX07582-20-UK MFLX07582-10-CH MFLX07582-10-IT MFLX07582-10-USS MFLX07582-00-AU MFLX07582-10-IL MFLX07582-00-EU MFLX07582-10
Masterflex® Modular Touch-Screen Dispensing Pump, Avantor®
Pompe Pompes péristaltiques
A versatile dispensing pump with an intuitive touch-screen interface for programming and control.
  • Flow rates from 0.001 to 1500 ml/min
  • Intuitive touch-screen interface
  • External connections on the back of the controller include phono jack connector for optional foot switch, RS-485 connections for daisy-chaining pumps, RS-232 and USB ports for connection
These versatile dispensing pumps are feature-rich and easy to program. Quickly set up single- or multiple-step programs depending on the requirements of your application. The controller will store up to 50 programs of up with 20 steps each; programs are e
Informations de livraison: Includes easy-Load II pump head(s), power adapter with 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.