Imprimer
MPC series plastic couplings
Fournisseur: CPC (Colder Products Company)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
212-0809EA 835 EUR
212-0809 212-0808 212-0807 212-0806 212-0805 212-0804 212-0814 212-0803 212-0802 212-0813 212-0812 212-0801 212-0811 212-0810
MPC series plastic couplings
Raccords et connecteurs
Couplings allow rapid disconnect and reconnect with one hand and without the need for tools.
Couplings are available in polysulphone (amber tint) or polycarbonate (purple tint) construction. Coupling inserts include seals constructed of clear silicone. All products meet USP Class VI criteria.
They feature an ergonomic thumb latch and an audible click when connected, assuring the user of a secure connection. Couplings can be rotated when connected, reducing the inconvenience of tube kinks. These items are sterilisable and reusable, yet economical enough to allow disposability.
Certifications: All products meet USP Class VI criteria and feature BSE-/TSE-free materials (ADCF).