Imprimer

Propargyl alcohol propoxylate 90%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): 1-(Prop-2-yn-1-yloxy)propan-2-ol

BD00857091-25G
BLDPBD00857091-25GEA 6 EUR
BLDPBD00857091-25G
Propargyl alcohol propoxylate 90%
Propargyl alcohol propoxylate

Storage condition: Sealed in dry, 2-8°C

Formule: C₆H₁₀O₂
Poids moléculaire: 114,142 g/mol
Température de stockage: Refrigerator
Numéro MDL: MFCD09751410
Numéro CAS: 3973-17-9

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR