Imprimer

4-(Morpholin-4-yl)-4-oxobut-2-enoic acid 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00875698-1G BD00875698-250 BD00875698-100
BLDPBD00875698-1GEA 244 EUR
BLDPBD00875698-1G BLDPBD00875698-250 BLDPBD00875698-100
4-(Morpholin-4-yl)-4-oxobut-2-enoic acid 98%
4-(Morpholin-4-yl)-4-oxobut-2-enoic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₈H₁₁NO₄
Poids moléculaire: 185,1772 g/mol
Température de stockage: Refrigerator
Numéro CAS: 90090-50-9

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR