Imprimer

N,N'-Bis(tert-butoxycarbonyl)-N,S-dimethylisothiourea 90%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): tert-Butyl N-[(1Z)-{[(tert-butoxy)carbonyl]imino}(methylsulphanyl)methyl]-N-methylcarbamate

BD00909996-1G BD00909996-250 BD00909996-100
BLDPBD00909996-1GEA 225 EUR
BLDPBD00909996-1G BLDPBD00909996-250 BLDPBD00909996-100
N,N'-Bis(tert-butoxycarbonyl)-N,S-dimethylisothiourea 90%
N,N'-Bis(tert-butoxycarbonyl)-N,S-dimethylisothiourea

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₁₃H₂₄N₂O₄S
Poids moléculaire: 304,406 g/mol
Température de stockage: Refrigerator
Numéro MDL: MFCD04973518
Numéro CAS: 215175-55-6

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR