Imprimer

(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): (S)-Methyl 6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride,Z-Lys-OMe·HCl

BD00935234-100 BD00935234-5G BD00935234-25G BD00935234-250 BD00935234-1G
BLDPBD00935234-100EA 32 EUR
BLDPBD00935234-100 BLDPBD00935234-5G BLDPBD00935234-25G BLDPBD00935234-250 BLDPBD00935234-1G
(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formule: C₁₅H₂₃ClN₂O₄
Poids moléculaire: 330,809 g/mol
Température de stockage: Refrigerator
Numéro MDL: MFCD00077025
Numéro CAS: 26348-68-5

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR