Imprimer

4-Bromo-8-methoxyisoquinoline 95%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00944693-250 BD00944693-100
BLDPBD00944693-250EA 433 EUR
BLDPBD00944693-250 BLDPBD00944693-100
4-Bromo-8-methoxyisoquinoline 95%
4-Bromo-8-methoxyisoquinoline

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₈BrNO
Poids moléculaire: 238,0800 g/mol
Température de stockage: Ambient
Numéro CAS: 1784377-21-4

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR