Imprimer

N,N-DIoctylacetamide 95%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00948639-250 BD00948639-1G
BLDPBD00948639-250EA 214 EUR
BLDPBD00948639-250 BLDPBD00948639-1G
N,N-DIoctylacetamide 95%
N,N-DIoctylacetamide

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₈H₃₇NO
Poids moléculaire: 283,4925 g/mol
Température de stockage: Ambient
Numéro MDL: MFCD30474888
Numéro CAS: 4088-41-9

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR