Imprimer

Ethyl 3-(thiazol-5-yl)acrylate 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00953092-1G BD00953092-100 BD00953092-250
BLDPBD00953092-1GEA 397 EUR
BLDPBD00953092-1G BLDPBD00953092-100 BLDPBD00953092-250
Ethyl 3-(thiazol-5-yl)acrylate 97%
Ethyl 3-(thiazol-5-yl)acrylate

Storage condition: Sealed in dry, 2-8°C

Formule: C₈H₉NO₂S
Poids moléculaire: 183,2276 g/mol
Température de stockage: Refrigerator
Numéro CAS: 1967005-05-5

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR