Imprimer

2-Methyl-4-(2,3,4,5-tetrahydro-1H-benzo[b]azepine-1-carbonyl)benzonitrile 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00957522-100 BD00957522-1G BD00957522-250
BLDPBD00957522-100EA 106 EUR
BLDPBD00957522-100 BLDPBD00957522-1G BLDPBD00957522-250
2-Methyl-4-(2,3,4,5-tetrahydro-1H-benzo[b]azepine-1-carbonyl)benzonitrile 97%
2-Methyl-4-(2,3,4,5-tetrahydro-1H-benzo[b]azepine-1-carbonyl)benzonitrile

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₉H₁₈N₂O
Poids moléculaire: 290,3590 g/mol
Température de stockage: Refrigerator
Numéro CAS: 335155-20-9

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR