Imprimer

9,9'-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-phenylene)bis(9H-carbazole) 98+%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00958149-250 BD00958149-5G BD00958149-1G BD00958149-100
BLDPBD00958149-250EA 57 EUR
BLDPBD00958149-250 BLDPBD00958149-5G BLDPBD00958149-1G BLDPBD00958149-100
9,9'-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-phenylene)bis(9H-carbazole) 98+%
9,9'-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-phenylene)bis(9H-carbazole)

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₃₆H₃₁BN₂O₂
Poids moléculaire: 534,4546 g/mol
Température de stockage: Ambient
Numéro CAS: 1082549-89-0

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR