Imprimer

1-Fluoro-4-(3-iodopropyl)benzene 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00971792-250
BLDPBD00971792-250EA 56 EUR
BLDPBD00971792-250
1-Fluoro-4-(3-iodopropyl)benzene 97%
1-Fluoro-4-(3-iodopropyl)benzene

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₉H₁₀FI
Poids moléculaire: 264,07 g/mol
Température de stockage: Refrigerator
Numéro CAS: 118156-84-6

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR