Imprimer

Ethyl 2-(bromotriphenyl-l5-phosphanyl)acetate 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD00977949-25G BD00977949-10G BD00977949-100 BD00977949-5G
BLDPBD00977949-25GEA 51 EUR
BLDPBD00977949-25G BLDPBD00977949-10G BLDPBD00977949-100 BLDPBD00977949-5G
Ethyl 2-(bromotriphenyl-l5-phosphanyl)acetate 97%
Ethyl 2-(bromotriphenyl-l5-phosphanyl)acetate

Storage condition: Inert atmosphere, Room Temperature

Formule: C₂₂H₂₂BrO₂P
Poids moléculaire: 429,2900 g/mol
Température de stockage: Ambient
Numéro CAS: 4083-85-6

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR