Imprimer

4-((6-Aminohexyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride 99%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01101120-1G BD01101120-250 BD01101120-100 BD01101120-50M
BLDPBD01101120-1GEA 1476 EUR
BLDPBD01101120-1G BLDPBD01101120-250 BLDPBD01101120-100 BLDPBD01101120-50M
4-((6-Aminohexyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride 99%
4-((6-Aminohexyl)oxy)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₉H₂₄ClN₃O₅
Poids moléculaire: 409,8640 g/mol
Température de stockage: Ambient
Numéro CAS: 2245697-88-3

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR