Imprimer

2-(3-Oxocyclopentyl)acetic acid 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01102247-250 BD01102247-100
BLDPBD01102247-250EA 302 EUR
BLDPBD01102247-250 BLDPBD01102247-100
2-(3-Oxocyclopentyl)acetic acid 98%
2-(3-Oxocyclopentyl)acetic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formule: C₇H₁₀O₃
Poids moléculaire: 142,1525 g/mol
Température de stockage: Freezer
Numéro MDL: MFCD00154377
Numéro CAS: 3128-05-0

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR