Imprimer

(S)-4-Hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98% + (99%ee)

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01104009-1G BD01104009-100 BD01104009-5G BD01104009-250
BLDPBD01104009-1GEA 558 EUR
BLDPBD01104009-1G BLDPBD01104009-100 BLDPBD01104009-5G BLDPBD01104009-250
(S)-4-Hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98% + (99%ee)
(S)-4-Hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆₈H₈₁O₄P
Poids moléculaire: 993,3421 g/mol
Température de stockage: Ambient
Numéro CAS: 1309446-14-7

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR