Imprimer

4,8-Bis(5-((2-ethylhexyl)thio)thiophen-2-yl)benzo[1,2-b:4,5-b']dithiophene 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01105686-100
BLDPBD01105686-100EA 64 EUR
BLDPBD01105686-100
4,8-Bis(5-((2-ethylhexyl)thio)thiophen-2-yl)benzo[1,2-b:4,5-b']dithiophene 98%
4,8-Bis(5-((2-ethylhexyl)thio)thiophen-2-yl)benzo[1,2-b:4,5-b']dithiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₃₄H₄₂S₆
Poids moléculaire: 643,0873 g/mol
Température de stockage: Ambient
Numéro CAS: 1613389-28-8

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR