Imprimer

2,5-Dipropoxyterephthalohydrazide 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01112456-100 BD01112456-1G BD01112456-5G BD01112456-250
BLDPBD01112456-100EA 47 EUR
BLDPBD01112456-100 BLDPBD01112456-1G BLDPBD01112456-5G BLDPBD01112456-250
2,5-Dipropoxyterephthalohydrazide 98%
2,5-Dipropoxyterephthalohydrazide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₄H₂₂N₄O₄
Poids moléculaire: 310,3489 g/mol
Température de stockage: Ambient
Numéro CAS: 2143456-42-0

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR