Imprimer

2-Oxo-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromene-3-carboxylic acid 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01117666-100 BD01117666-1G BD01117666-250
BLDPBD01117666-100EA 184 EUR
BLDPBD01117666-100 BLDPBD01117666-1G BLDPBD01117666-250
2-Oxo-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromene-3-carboxylic acid 98%
2-Oxo-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromene-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₆H₁₇BO₆
Poids moléculaire: 316,1136 g/mol
Température de stockage: Refrigerator
Numéro CAS: 1442461-73-5

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR