Imprimer

2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3,9-dihydro-6H-purine-6-thione hydrate 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01119426-100 BD01119426-5G BD01119426-1G BD01119426-250
BLDPBD01119426-100EA 53 EUR
BLDPBD01119426-100 BLDPBD01119426-5G BLDPBD01119426-1G BLDPBD01119426-250
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3,9-dihydro-6H-purine-6-thione hydrate 98%
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3,9-dihydro-6H-purine-6-thione hydrate

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formule: C₁₀H₁₅N₅O₅S
Poids moléculaire: 317,3216 g/mol
Température de stockage: Freezer
Numéro MDL: MFCD00149353
Numéro CAS: 345909-25-3

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR