Imprimer

4-Hexyl-4H-dithieno[3,2-b:2',3'-d]pyrrole 98%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01124262-250 BD01124262-1G BD01124262-100
BLDPBD01124262-250EA 45 EUR
BLDPBD01124262-250 BLDPBD01124262-1G BLDPBD01124262-100
4-Hexyl-4H-dithieno[3,2-b:2',3'-d]pyrrole 98%
4-Hexyl-4H-dithieno[3,2-b:2',3'-d]pyrrole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₁₄H₁₇NS₂
Poids moléculaire: 263,4215 g/mol
Température de stockage: Ambient
Numéro CAS: 427875-87-4

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR