Imprimer

(E)-1-(Pyridin-2-yl)ethanone O-benzyl oxime 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BD01125021-1G BD01125021-100 BD01125021-250
BLDPBD01125021-1GEA 288 EUR
BLDPBD01125021-1G BLDPBD01125021-100 BLDPBD01125021-250
(E)-1-(Pyridin-2-yl)ethanone O-benzyl oxime 97%
(E)-1-(Pyridin-2-yl)ethanone O-benzyl oxime

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₄H₁₄N₂O
Poids moléculaire: 226,2738 g/mol
Température de stockage: Refrigerator
Numéro CAS: 216753-06-9

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR